Learn More
LIGHTHOUSE WORLDWIDE SOLUTIONS INC SOLAIR 3100 BUNDLE
SDP
Catalog No. NC3761486
Encompass
  • Versatile application for cleanroom certification to ISO 14644-1, EU GMP Annex 1, and FS-209E standards
  • Capable of compressed gas testing down to 0.3 µm particle size
  • Integrated thermal printer for immediate results reporting
  • Supports monitoring of environmental parameters like temperature and humidity via optional sensors
  • Intuitive operation with a 5.7 in color TFT touch screen
  • Durable stainless steel construction with over 20-year lifespan
  • Stores up to 3,000 particle and environmental data records
  • 21 CFR Part 11 compliant secure data transfer to USB flash drive
  • Flow rate of 1.0 CFM (28.3 LPM)
  • Particle size range from 0.3 to 25.0 µm
  • Up to 8 particle channels
  • Multi-language support including English and Spanish
Catalog No. NC3761486 Supplier LIGHTHOUSE WORLDWIDE SOLUTIONS INC Supplier No. 4029994501
Add to Cart
Add to Cart
Request a Quote
  • Versatile application for cleanroom certification to ISO 14644-1, EU GMP Annex 1, and FS-209E standards
  • Capable of compressed gas testing down to 0.3 µm particle size
  • Integrated thermal printer for immediate results reporting
  • Supports monitoring of environmental parameters like temperature and humidity via optional sensors
  • Intuitive operation with a 5.7 in color TFT touch screen
  • Durable stainless steel construction with over 20-year lifespan
  • Stores up to 3,000 particle and environmental data records
  • 21 CFR Part 11 compliant secure data transfer to USB flash drive
  • Flow rate of 1.0 CFM (28.3 LPM)
  • Particle size range from 0.3 to 25.0 µm
  • Up to 8 particle channels
  • Multi-language support including English and Spanish