Learn More
TARGETMOL CHEMICALS INC SOLAMARGINE 5MG
SDP
Catalog No. 502576824
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Solamargine ((delta)-Solanigrine) is a major steroidal alkaloid glycoside extracted from a traditional Chinese medicine herb Solanum nigrum L. (SNL); has been shown to inhibit growth and induce apoptosis of various cancer cells. purity: 99%
Catalog No. 50-257-6824 Supplier TARGETMOL CHEMICALS INC Supplier No. T40345MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Solamargine ((delta)-Solanigrine) is a major steroidal alkaloid glycoside extracted from a traditional Chinese medicine herb Solanum nigrum L. (SNL); has been shown to inhibit growth and induce apoptosis of various cancer cells. purity: 99%