Learn More
Cayman Chemical Solamargn 10mg
SDP
Catalog No. 502941820
Encompass_Preferred
A glycoalkaloid with anticancer activity; decreases viability of H1650, H1975, PC-9, A549, and H1299 NSCLC cells at 6 µM; reduces tumor growth in an A549 mouse xenograft model at 8 mg/kg per day
Catalog No. 50-294-1820 Supplier Cayman Chemical Supplier No. 2811010 MG
A glycoalkaloid with anticancer activity; decreases viability of H1650, H1975, PC-9, A549, and H1299 NSCLC cells at 6 µM; reduces tumor growth in an A549 mouse xenograft model at 8 mg/kg per day