Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana C. diff Startup Kit
SDP

Catalog No. 23043120 Shop All QuidelOrtho Products
Change view
Click to view available options
Test Time:
<45 min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Test Time
23-043-120 <45 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-120 Supplier Quidel Supplier No. M923
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Quidel Solana C. difficile Assay is available with Virena, near real-time surveillance information to provide assessment of incidence throughout a health system to monitor C. diff infections.

The Solana C. diff test has a fast TAT, power of Virena™, and ability to batch test up to 12 tests at a time. Solana provides a 93% sensitivity and 99% specificity rate with an inhibition rate of 0.2%
  • CLIA-Moderate Molecular Assay with accurate molecular results
  • Turnaround time in under 45 minutes, multiplexing up to 12 tests per run
  • Access to the Power of Virena (Surveillance)
  • LIS/EMR Connectivity

Specifications

Sensitivity 93%
Type C. difficile Test
For Use With (Application) Monitor C. diff infections
Includes 1 leased Solana Instrument, C. diff test kit and controls, 95C Lysis Heat Block, Printer, Labels, Tray, 3 Racks & Stylus
Sample Size 12 Tests
Specificity 0.99
Test Time <45 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.