Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana™ Flu Assay Startup Kit
GSA_VASDP

Catalog No. 23043107 Shop All QuidelOrtho Products
Change view
Click to view available options
Type:
Influenza A and B Assay Kit
Molecular Diagnostic Instrument
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-043-107 Influenza A and B Assay Kit
23-043-106 Molecular Diagnostic Instrument
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-043-107 Supplier Quidel Supplier No. M300
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Process Buffer, Reaction Tubes

For rapid differential diagnosis of acute influenza A and influenza B viral infections.

  • CLIA Complexity: Moderate
  • Detection and differentiation of Influenza A and Influenza B
  • Rapid qualitative in vitro diagnostic test for the detection and differentiation of influenza A and influenza B viral RNA.
  • Sample collected from nasal and nasopharyngeal swabs from patients with signs and symptoms of respiratory infection.
  • HDA Technology
  • No need for culture confirmation




Order Info

48 tests/Pk.

Recommended Storage

2 to 8°C

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
Control Sets M106
CPT Code 87502
CE Marker Yes
Clia Complexity Moderate
Detectable Analytes Influenza A and B
Sensitivity 98.6.%/100.0% (Culture), 97.2%/100% (PCR)
Type Influenza A and B Assay Kit
For Use With (Application) For diagnosis of Influenza A&B
Includes Process Buffer, Reaction Tubes
Quantity 48/Pk.
Specificity 95.1.% / 99.3% (culture)  96.8%/98.9% (PCR)
Test Time ≈45 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.