Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana™ GAS Assay
GSA_VASDP

Catalog No. 23043116 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
48 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-116 48 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-116 Supplier Quidel Supplier No. M801

Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher HealthCare Account Representative for more information.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Kit components: Dilution Buffer (48 tubes/kit 0.5mL), Lysis Buffer ((48 tubes/kit 0.5mL), Reaction Tubes

Amplifies and detects GAS DNA present in throat swab specimens obtained from symptomatic patients. Quidel™ Solana™ GAS Assay is rapid in vitro diagnostic test for the qualitative detection of Streptococcus pyogenes.

  • Intended for use only with Solana instrument
  • Assay when performed on Solana instrument, allows for rapid, accurate detection of Group A Strep without need for culture confirmation
  • Features HDA technology that acts as rapid method of nucleic acid amplification that does not require thermal cycler
  • Contains Lyophilized Master Mix reagent that is is easy-to-use format, add processed sample to rehydrate
  • Small, easy-to-use instrument
  • CLIA moderately complex feature requires minimal testing
  • No freezer needed, 2° to 8°C storage
  • Shelf life: 12 months

Order Info

Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage Lysis, Dilution and Reaction Tubes; 2° to 8°C
Detectable Analytes Streptococci (Group A)
Format Kit
Sample Type Throat swab specimens obtained from patients symptomatic of acute pharyngitis
Sensitivity 98.2% (95% CI 95.5% to 99.3%)
Type GAS Assay
For Use With (Application) Qualitative detection of Group A β-hemolytic Streptococcus (Streptococcus pyogenes) nucleic acids isolated from throat swab specimens obtained from patients with signs and symptoms of pharyngitis, such as sore throat.
Includes Kit components: Dilution Buffer (48 tubes/kit 0.5mL), Lysis Buffer ((48 tubes/kit 0.5mL), Reaction Tubes
Organism Group Streptococcus
Quantity 48 Tests/Kit
Specificity 97.2% (95% CI 95.9% to 98.1%)
Test Time ≈35 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.