Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana GBS Assay Kit
SDP

Catalog No. 23043128 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
48
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-128 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-128 Supplier Quidel Supplier No. M311
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Solana GBS Assay is an in vitro diagnostic test for the direct, qualitative detection of Group B Streptococcus from vaginal/rectal swabs following 18 to 24 hours incubation in LIM or Carrot broth cultures.

  • HDA technology - Rapid method of isothermal nucleic acid amplification that does not require thermal cycler
  • Lyophilized Master Mix reagent - Easy-to-use format, just add processed sample to rehydrate
  • Small, easy-to-use instrument - Eliminates the need for dedicated molecular space and costly capital equipment, allowing for testing in smaller labs
  • CLIA moderately complex - Requires minimal training
  • Refrigerated storage - No freezer needed, 2˚C to 8˚C storage
  • Long shelf life - Currently 12 months from date of manufacture
  • Room temperature at set up - No ice or cooling block required

Specifications

Content And Storage 2°C to 8°C
Sample Type Vaginal/rectal swab specimens cultured for 18 to 24 hours in either LIM or Carrot Broth
Includes Process Buffer 48 tubes/kit, Reaction Tubes 48 tubes/kit
Quantity 48
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.