Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana™ Group A Streptococcus Test Kit
SDP

Catalog No. 23043100 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
48/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-100 48/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Refer to Package Insert for list of warnings
Catalog No. 23-043-100 Supplier Quidel Supplier No. M301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Refer to Package Insert for list of warnings

Includes

  • Test Kit:
  • Package insert
  • Dilution Buffer
  • Lysis Buffer
  • Reaction Tubes

in vitro diagnostic test for the qualitative detection of Group A β-hemolytic Streptococcus (Streptococcus pyogenes) nucleic acids. The Solana GAS Assay is for use with samples isolated from throat swab specimens from patients with signs and symptoms of pharyngitis, such as sore throat.

  • HDA Technology
  • CLIA Moderate
  • No need for Culture Confirmation
  • Time to result: Approximately 30 to 35 minutes
  • Lyophilized Master Mix reagent
  • No ice or cooling block required

Certifications

CLIA-Moderate

Specifications

Content And Storage 2°C to 8°C
Clia Complexity Moderate
Sample Type Throat Swab
Sensitivity 98.2%
Type Group A Strep Test
For Use With (Application) For diagnosis of Group A Streptococcus
Quantity 48/Pk.
Specificity 0.972
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.