Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana HSV 1+2/VZV Control Set
SDP

Catalog No. 23043115 Shop All QuidelOrtho Products
Change view
Click to view available options
Content And Storage:
2°C to 8°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
23-043-115 2°C to 8°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-115 Supplier Quidel Supplier No. M118
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quidel Solana HSV 1+2/VZV Control Set for aid in diagnosis of Herpes Simplex Virus (HSV) 1 and 2 and Varicella-zoster virus (VZV).

  • Used in conjunction with Quidel Solana HSV 1 and 2/VZV Test Kit
  • Storage: 2-8°C
  • CLIA Complexity: Moderate - Prescription Use Only

Specifications

Content And Storage 2°C to 8°C
Clia Complexity Moderate (Prescription Use Only)
Type Solana HSV 1+2/VZV Control Set
Includes Process buffer, Reaction tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.