Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana HSV 1+2/VZV Starter Kit
SDP

Catalog No. 23043113 Shop All QuidelOrtho Products
Change view
Click to view available options
Test Time:
≈50 min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Test Time
23-043-113 ≈50 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-113 Supplier Quidel Supplier No. M919
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quidel™ Solana HSV 1+2/VZV Assay is multiplex in vitro diagnostic test for detection and differentiation of Herpes simplex virus type 1 (HSV-1), Herpes simplex virus type 2 (HSV-2), and Varicella-zoster virus (VZV) nucleic acids isolated and purified from cutaneous or mucocutaneous swab specimens.

  • For detection and differentiation of Herpes Simplex Virus (HSV) 1 and 2 and Varicella-zoster virus (VZV)/li>
  • HDA technology: Rapid method of nucleic acid amplification, does not require thermal cycler/li>
  • Lyophilized Master Mix reagent: Easy-to-use format, add processed sample to rehydrate/li>
  • CLIA complex: Moderate - Prescription Use Only
  • Refrigerated storage: No freezer needed/li>
  • Eliminates need for dedicated molecular space and capital equipment, allowing for testing in smaller labs/li>

Order Info

Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher HealthCare Account Representative for more information

Specifications

Content And Storage 2°C to 8°C (No ice or cooling block required)
Clia Complexity Moderate (Prescription Use Only)
Sample Type Cutaneous or mucocutaneous lesion samples obtained from symptomatic patients
Sensitivity Cutaneous: 100% (HSV-1), 92.3% (HSV-2), 100% (VZV); Mucocutaneous: 100% (HSV-1), 99.1% (HSV-2), 100% (VZV); Non categorized: 100% (HSV-1), 100% (HSV-2), 100% (VZV)
Type Solana HSV 1+2/VZV Starter Kit
For Use With (Application) Diagnosis of Herpes Simplex Virus (HSV) 1 and 2 and Varicella-zoster virus (VZV)
Includes One leased Solana Instrument, HSV 1+2/VZV Assay Kit (48 tests), Solana HSV 1+2/VZV Control Set, 95C Lysis heat block, Printer, Labels, Tray, three racks and stylus; Lyophilized Master Mix reagent
Specificity Cutaneous: 98% (HSV-1), 94.4% (HSV-2), 96.5% (VZV); Mucocutaneous: 96.4% (HSV-1), 97.2% (HSV-2), 98.6% (VZV); Non categorized: 97.9% (HSV-1), 96.6% (HSV-2), 94.1% (VZV)
Test Time ≈50 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.