Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana HSV 1+2/VZV Test Kit
GSA_VASDP

Catalog No. 23043114 Shop All QuidelOrtho Products
Change view
Click to view available options
Test Time:
≈50 min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Test Time
23-043-114 ≈50 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-114 Supplier Quidel Supplier No. M302
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quidel™ Solana HSV 1+2/VZV Assay is multiplex in vitro diagnostic test for detection and differentiation of Herpes simplex virus type 1 (HSV-1), Herpes simplex virus type 2 (HSV-2), and Varicella-zoster virus (VZV) nucleic acids isolated and purified from cutaneous or mucocutaneous swab specimens.

  • For detection and differentiation of Herpes Simplex Virus (HSV) 1 and 2 and Varicella-zoster virus (VZV)
  • HDA technology: Rapid method of nucleic acid amplification, does not require thermal cycler
  • Lyophilized Master Mix reagent: Easy-to-use format, add processed sample to rehydrate
  • CLIA complex: Moderate - Prescription Use Only
  • Refrigerated storage: No freezer needed
  • Eliminates need for dedicated molecular space and capital equipment, allowing for testing in smaller labs

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C (No ice or cooling block required)
Clia Complexity Moderate (Prescription Use Only)
Sample Type Cutaneous or mucocutaneous lesion samples obtained from symptomatic patients
Sensitivity Cutaneous: 100% (HSV-1), 92.3% (HSV-2), 100% (VZV); Mucocutaneous: 100% (HSV-1), 99.1% (HSV-2), 100% (VZV); Non categorized: 100% (HSV-1), 100% (HSV-2), 100% (VZV)
Type Solana HSV 1+2/VZV Starter Kit
For Use With (Application) Diagnosis of Herpes Simplex Virus (HSV) 1 and 2 and Varicella-zoster virus (VZV)
Includes Package insert, Dilution buffer, Lysis buffer, Reaction tubes; Lyophilized Master Mix reagent
Specificity Cutaneous: 98% (HSV-1), 94.4% (HSV-2), 96.5% (VZV); Mucocutaneous: 96.4% (HSV-1), 97.2% (HSV-2), 98.6% (VZV); Non categorized: 97.9% (HSV-1), 96.6% (HSV-2), 94.1% (VZV)
Test Time ≈50 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.