Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana SARS-CoV-2 Assay
SDP

Catalog No. 23044159 Shop All QuidelOrtho Products
Change view
Click to view available options
Type:
Nasopharyngeal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-044-159 Nasopharyngeal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-044-159 Supplier Quidel Supplier No. M956

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isothermal Reverse Transcriptase-Helicase-Dependent Amplification (RT-HDA) assay intended for the qualitative detection of nucleic acid from SARS-CoV-2 in nasopharyngeal & nasal swab specimens from individuals suspected of COVID-19 by their healthcare provider.

  • RT-HDA technology
  • Frozen Master Mix
  • Small, easy-to-use instrument
  • CLIA moderately complex
  • Storage (Process Buffer 2°C to 8°C storage, Master Mix -70°C or below)
  • Room temperature set up

Specifications

Certifications/Compliance Emergency Use Authorization (EUA)
Type Nasopharyngeal
For Use With (Application) Solana Instrument
Includes Universal Transport Media
Quantity 96/Kit
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.