Learn More
Quidel Solana SARS-COV-2 Negative Control
SDP
Catalog No. 23044164 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
1 Tube/Kit
Catalog No. Quantity
23-044-164 1 Tube/Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-044-164 Supplier Quidel Supplier No. M5203
Add to Cart
Add to Cart
This item is not returnable. View return policy
Included with kit, additional may be ordered separately
  • 1 tube per kit
  • 2 mL
  • 2-8°C Storage
Specifications
Description Negative Control
Concentration 2 mL
Detectable Analytes SARS-CoV-2
Technology Reverse Transcriptase Assay Control
Type Quality Control
Quantity 1 Tube/Kit