Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana Trichomonas Starter Kit
SDP

Catalog No. 23043105 Shop All QuidelOrtho Products
Change view
Click to view available options
Type:
Assay
Starter Kit
Sample Type:
Urine
Vaginal Swab
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Sample Type
23-043-105 Starter Kit Vaginal Swab
23-043-104 Starter Kit Urine
23-043-103 Assay Urine
23-043-102 Assay Vaginal Swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-043-105 Supplier Quidel Supplier No. M914
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Delivers molecular results faster with bench top testing system. Quidel™ Solana™ Instrument with Trichomonas Urine Test Kit uses helicase-dependent amplification (HDA) with fluorescence detection

  • Molecular Biology grade. D
  • pH of solution at 25°C: 6.85-7.15
  • Protease: None detected
  • RNase: None detected

  • Bench top instrument
  • Helicase-dependent amplification (HDA) with fluorescence detection
  • Capable of testing a single specimen or batching up to 12 tests at a time
  • Intuitive, touch-screen interface with guided operation and customizable settings
  • Isothermal amplification ensures molecular sensitivity with moderate complexity
  • Eliminates thermal cycling to aid in faster time to result (as little as 30 minutes)
  • Simplified workflow prevents the need for specialized molecular operator training, making Solana easily adoptable
  • Small foot-print facilitates decentralization of the instruments to bring them closer to the patient
  • Barcode technology reduces time and errors associated with manually recording patient and operator ID
  • Reduce time manually recording test results and risk of transcription errors
  • Connect to many of existing information systems
  • Comes with power of Virena, Quidel's proprietary wireless data management and surveillance ecosystem

Order Info

Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher HealthCare Account Representative for more information.

Specifications

Description Molecular Instrument
Clia Complexity Moderate
Sample Type Vaginal Swab
Type Starter Kit
For Use With (Application) Detection of Trichomonas from vaginal swab
Includes Instrument, Heat Block, Printer, Labels, Tray, Racks and Stylus
Memory Test, QC, and Calibration results stored on-board
Quantity 48 Tests/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.