Learn More
Medchemexpress LLC Solanesol | 13190-97-1 | 631.07 | 100 MG
SDP
Catalog No. 5000389656
Encompass_Preferred
Solanesol is an orally active aliphatic terpene alcohol primarily found in tobacco and other Solanaceae plants. It induces HO-1 and Hsp70 expression, activates p38 and Akt signaling pathways, and inhibits Apoptosis by reducing caspase-3 and PARP cleavage. Solanesol exhibits antioxidant, anti-inflammatory, and neuroprotective activities.
  • Suitable for research in Huntington's disease, alcoholic liver disease, chronic inflammatory pain, anxiety, Alzheimer's disease, and bipolar disorder.
  • Induces HO-1 and Hsp70 expression.
  • Activates p38 and Akt signaling pathways.
  • Inhibits apoptosis.
  • Possesses antioxidant, anti-inflammatory, and neuroprotective properties.
Catalog No. 50-003-89656 Supplier Medchemexpress LLC Supplier No. HYN0576100MG
Solanesol is an orally active aliphatic terpene alcohol primarily found in tobacco and other Solanaceae plants. It induces HO-1 and Hsp70 expression, activates p38 and Akt signaling pathways, and inhibits Apoptosis by reducing caspase-3 and PARP cleavage. Solanesol exhibits antioxidant, anti-inflammatory, and neuroprotective activities.
  • Suitable for research in Huntington's disease, alcoholic liver disease, chronic inflammatory pain, anxiety, Alzheimer's disease, and bipolar disorder.
  • Induces HO-1 and Hsp70 expression.
  • Activates p38 and Akt signaling pathways.
  • Inhibits apoptosis.
  • Possesses antioxidant, anti-inflammatory, and neuroprotective properties.