Learn More
Apexbio Technology LLC Solanesol 13190-97-1 200mg
SDP
Catalog No. 5000064069
Encompass_Preferred
Solanesol is a lipophilic terpene alcohol derived from plants in the Solanaceae family characterized by anti-inflammatory neuroprotective and antimicrobial activity In vitro studies indicate solanesol regulates specific signaling pathways implicated in inflammatory responses and neurodegenerative processes possibly through modulation of enzyme activity and intracellular targets Experimental evidence demonstrates solanesol inhibits inflammation-associated enzymes with reported IC50 values typically ranging from low to mid micromolar concentrations depending on cell types and assay conditions This compound is utilized in biochemical research focused on elucidating molecular pathways involved in inflammation-mediated diseases and evaluating therapeutic potential as a neuroprotective agent
Catalog No. 50-000-64069 Supplier Apexbio Technology LLC Supplier No. B8776200
Solanesol is a lipophilic terpene alcohol derived from plants in the Solanaceae family characterized by anti-inflammatory neuroprotective and antimicrobial activity In vitro studies indicate solanesol regulates specific signaling pathways implicated in inflammatory responses and neurodegenerative processes possibly through modulation of enzyme activity and intracellular targets Experimental evidence demonstrates solanesol inhibits inflammation-associated enzymes with reported IC50 values typically ranging from low to mid micromolar concentrations depending on cell types and assay conditions This compound is utilized in biochemical research focused on elucidating molecular pathways involved in inflammation-mediated diseases and evaluating therapeutic potential as a neuroprotective agent