Learn More
Selleck Chemical LLC Solanesol (Nonaisoprenol) S2360-100mg
SDP
Catalog No. 503112376
Encompass_Preferred
Solanesol (Nonaisoprenol Betulanonaprenol) is a long-chain polyisoprenoid alcohol compound with inhibitory activities toward FAK It mainly accumulates in solanaceous crops including tobacco tomato potato eggplant and pepper plants
Catalog No. 50-311-2376 Supplier Selleck Chemical LLC Supplier No. S2360100MG
Solanesol (Nonaisoprenol Betulanonaprenol) is a long-chain polyisoprenoid alcohol compound with inhibitory activities toward FAK It mainly accumulates in solanaceous crops including tobacco tomato potato eggplant and pepper plants