Learn More
Medchemexpress LLC Solangepras | 2254706-21-1 | 99.7% | C24H29F2N5O3 | 10MG
SDP
Catalog No. 5000259764
Encompass_Preferred
Solangepras (CVN424) is an orally active, brain-penetrant inverse agonist of the orphan G protein-coupled receptor GPR6, developed for preclinical Parkinson's disease research. It exhibits high binding affinity and potent functional activity in cellular assays and is supplied as a solid research compound for laboratory studies.
  • Orally active and brain-penetrant small molecule suitable for in vivo studies.
  • High affinity GPR6 inverse agonist (Ki ≈ 9.4 nM).
  • Potent functional activity (EC50 ≈ 38 nM) in relevant assays.
  • Available as a stable solid for bench storage and formulation.
  • Provided at high chemical purity for research use.
Catalog No. 50-002-59764 Supplier Medchemexpress LLC Supplier No. HY134661A10MG
Solangepras (CVN424) is an orally active, brain-penetrant inverse agonist of the orphan G protein-coupled receptor GPR6, developed for preclinical Parkinson's disease research. It exhibits high binding affinity and potent functional activity in cellular assays and is supplied as a solid research compound for laboratory studies.
  • Orally active and brain-penetrant small molecule suitable for in vivo studies.
  • High affinity GPR6 inverse agonist (Ki ≈ 9.4 nM).
  • Potent functional activity (EC50 ≈ 38 nM) in relevant assays.
  • Available as a stable solid for bench storage and formulation.
  • Provided at high chemical purity for research use.