Learn More
Medchemexpress LLC HY-N7271 1mg , Solanidine CAS:80-78-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN72711MG
Encompass_Preferred
HY-N7271 1mg Solanidine CAS: 80-78-4 Solanidine is a cholestane alkaloid isolated from several potato species including Solanum demissum, Solanum acaule, and Solanum tuberosum. Solanidine can inhibit proliferation and exhibit obvious antitumor effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1971