Learn More
Bioworld Solanum tuberosum (Potato) Lectin (STA/STL) - Rhodamine (TRITC)
Catalog No. 50199164
Encompass
Technical Specifications:Ligand Source:Solanum tuberosumMolecular Weight:100 kDaCarbohydrate Specificity:(GlcNAcβ4)nInhibiting/Eluting Sugar:Chitin hydrolysateCarbohydrate for Elution:0.2MChitin hydrolysateBlood Group Specificity:N/AConjugate: RhodamineForm:LiquidIsoelectric (pI):>9Storage: -20°CApplication note: N/A
Catalog No. 50-199-164 Supplier Bioworld Supplier No. 217610551
Technical Specifications:Ligand Source:Solanum tuberosumMolecular Weight:100 kDaCarbohydrate Specificity:(GlcNAcβ4)nInhibiting/Eluting Sugar:Chitin hydrolysateCarbohydrate for Elution:0.2MChitin hydrolysateBlood Group Specificity:N/AConjugate: RhodamineForm:LiquidIsoelectric (pI):>9Storage: -20°CApplication note: N/A