Learn More
Grainger Solar Fuse 125A PVS-R Series 600VDC
Catalog No. 502749841
Encompass_Preferred
The Bussmann 14L662 is a 125 amp solar fuse that provides overcurrent protection for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass endcaps and zinc-plated steel blade terminals. This UL and CSA listed Class T fuse has a high interrupting rating of 200kA at 1000V DC. With dimensions of 1-1/4"L x 1/2"W, this fuse enables safe, reliable protection against overloads and short circuits in residential, commercial, and utility-scale solar applications.
Catalog No. 50-274-9841 Supplier Grainger Supplier No. 14L662
Add to Cart
Add to Cart
The Bussmann 14L662 is a 125 amp solar fuse that provides overcurrent protection for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass endcaps and zinc-plated steel blade terminals. This UL and CSA listed Class T fuse has a high interrupting rating of 200kA at 1000V DC. With dimensions of 1-1/4"L x 1/2"W, this fuse enables safe, reliable protection against overloads and short circuits in residential, commercial, and utility-scale solar applications.