Learn More
Grainger Solar Fuse 150A PVS-R Series 600VDC
Catalog No. 502749456
Encompass_Preferred
The BUSSMANN Solar Fuse 150 Amp is a UL listed, current limiting, high interrupting rating, finger safe solar fuse. It has a voltage rating of 1000VDC and a current rating of 150A. This fuse features a clear housing for easy blown fuse identification. It has nickel plated brass end caps and is finger safe with a dimension of 1-1/4"L x 1-1/4"W x 1-3/4"H. This fuse provides overcurrent protection for photovoltaic systems and solar panel arrays.
Catalog No. 50-274-9456 Supplier Grainger Supplier No. 14L663
The BUSSMANN Solar Fuse 150 Amp is a UL listed, current limiting, high interrupting rating, finger safe solar fuse. It has a voltage rating of 1000VDC and a current rating of 150A. This fuse features a clear housing for easy blown fuse identification. It has nickel plated brass end caps and is finger safe with a dimension of 1-1/4"L x 1-1/4"W x 1-3/4"H. This fuse provides overcurrent protection for photovoltaic systems and solar panel arrays.