Learn More
Grainger Solar Fuse 200A PVS-R Series 600VDC
Catalog No. 502749694
Encompass_Preferred
The Bussmann Solar Fuse 200 Amp is a 200 amp solar fuse designed for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass end caps and copper terminals. This fast-acting Class T fuse provides overcurrent protection for PV source circuits, PV output circuits, PV combiner boxes, and PV disconnects. It has a compact 13/32" x 1-1/2" body size and includes a blown fuse indicator for easy troubleshooting. The 200 amp rating allows it to be used on high current PV systems.
Catalog No. 50-274-9694 Supplier Grainger Supplier No. 14L665
The Bussmann Solar Fuse 200 Amp is a 200 amp solar fuse designed for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass end caps and copper terminals. This fast-acting Class T fuse provides overcurrent protection for PV source circuits, PV output circuits, PV combiner boxes, and PV disconnects. It has a compact 13/32" x 1-1/2" body size and includes a blown fuse indicator for easy troubleshooting. The 200 amp rating allows it to be used on high current PV systems.