Learn More
Grainger Solar Fuse 20A PVS-R Series 600VDC
Catalog No. 502752716
Encompass_Preferred
The Grainger Approved Solar Fuse is a 15 amp, 250 volt, time-delay fuse designed for solar panel applications. It features a ceramic body construction and measures 1-1/4" long x 1/4" diameter. The fuse helps protect solar panels and inverters from overcurrent and short circuit conditions. It has a time-delay feature to allow harmless inrush currents during start up. The fuse body color coding makes it easy to identify the amperage rating. This fuse provides overcurrent protection in DC photovoltaic systems up to 600 volts.
Catalog No. 50-275-2716 Supplier Grainger Supplier No. 14L651
Add to Cart
Add to Cart
The Grainger Approved Solar Fuse is a 15 amp, 250 volt, time-delay fuse designed for solar panel applications. It features a ceramic body construction and measures 1-1/4" long x 1/4" diameter. The fuse helps protect solar panels and inverters from overcurrent and short circuit conditions. It has a time-delay feature to allow harmless inrush currents during start up. The fuse body color coding makes it easy to identify the amperage rating. This fuse provides overcurrent protection in DC photovoltaic systems up to 600 volts.