Learn More
Grainger Solar Fuse 225A PVS-R Series 600VDC
Catalog No. 502752083
Encompass_Preferred
The Grainger Approved Solar Fuse is a 15 amp, 250 volt, time-delay fuse designed for solar panel applications. It features a ceramic body construction and measures 1-1/4" long x 1/4" diameter. The fuse has radial leads and includes a 5 amp interrupting rating to provide short circuit protection. This fuse helps protect solar panels, inverters, and other DC equipment in photovoltaic systems. It can handle up to 1500 volts DC. The time-delay feature allows harmless inrush currents while preventing nuisance opening from temporary overloads.
Catalog No. 50-275-2083 Supplier Grainger Supplier No. 14L666
The Grainger Approved Solar Fuse is a 15 amp, 250 volt, time-delay fuse designed for solar panel applications. It features a ceramic body construction and measures 1-1/4" long x 1/4" diameter. The fuse has radial leads and includes a 5 amp interrupting rating to provide short circuit protection. This fuse helps protect solar panels, inverters, and other DC equipment in photovoltaic systems. It can handle up to 1500 volts DC. The time-delay feature allows harmless inrush currents while preventing nuisance opening from temporary overloads.