Learn More
Grainger Solar Fuse 250A PVS-R Series 600VDC
Catalog No. 502749849
Encompass_Preferred
The Bussmann Solar Fuse 250 Amps is a 250 amp solar fuse designed for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass end caps and blade terminals. This fast-acting Class T fuse provides overcurrent protection for PV source circuits, PV output circuits, and PV combiner boxes. It has a compact 13/32" x 1-1/2" body size and includes a blown fuse indicator for easy troubleshooting. The Bussmann 14L667 model is UL and CSA recognized for PV applications.
Catalog No. 50-274-9849 Supplier Grainger Supplier No. 14L667
Add to Cart
Add to Cart
The Bussmann Solar Fuse 250 Amps is a 250 amp solar fuse designed for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass end caps and blade terminals. This fast-acting Class T fuse provides overcurrent protection for PV source circuits, PV output circuits, and PV combiner boxes. It has a compact 13/32" x 1-1/2" body size and includes a blown fuse indicator for easy troubleshooting. The Bussmann 14L667 model is UL and CSA recognized for PV applications.