Learn More
Grainger Solar Fuse 25A PVS-R Series 600VDC
Catalog No. 502749580
Encompass_Preferred
The Bussmann 14L652 is a 25 amp solar fuse that provides overcurrent protection for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass endcaps and zinc-plated steel blades. This UL and CSA listed Class T fuse has a fast-acting, high interrupting rating to minimize damage to PV components. With dimensions of 1-1/4"L x 1/4"W, this fuse can be used in in-line fuse holders or fuse blocks. The 14L652 solar fuse is ideal for protecting PV source circuits, PV output circuits, PV combiner boxes, and other DC applications up to 25 amps in residential or commercial systems.
Catalog No. 50-274-9580 Supplier Grainger Supplier No. 14L652
The Bussmann 14L652 is a 25 amp solar fuse that provides overcurrent protection for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass endcaps and zinc-plated steel blades. This UL and CSA listed Class T fuse has a fast-acting, high interrupting rating to minimize damage to PV components. With dimensions of 1-1/4"L x 1/4"W, this fuse can be used in in-line fuse holders or fuse blocks. The 14L652 solar fuse is ideal for protecting PV source circuits, PV output circuits, PV combiner boxes, and other DC applications up to 25 amps in residential or commercial systems.