Learn More
Grainger Solar Fuse 300A PVS-R Series 600VDC
Catalog No. 502752084
Encompass_Preferred
The BUSSMANN Solar Fuse 300 Amps is a 300 amp solar fuse designed for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass end caps and blade terminals. This fast-acting Class T fuse provides overcurrent protection for PV source circuits, PV output circuits, and PV combiner boxes. It has a compact 13/32" x 1-1/2" body size and includes a blown fuse indicator for easy troubleshooting. The BUSSMANN Solar Fuse line is UL and CSA recognized for PV systems.
Catalog No. 50-275-2084 Supplier Grainger Supplier No. 14L668
The BUSSMANN Solar Fuse 300 Amps is a 300 amp solar fuse designed for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass end caps and blade terminals. This fast-acting Class T fuse provides overcurrent protection for PV source circuits, PV output circuits, and PV combiner boxes. It has a compact 13/32" x 1-1/2" body size and includes a blown fuse indicator for easy troubleshooting. The BUSSMANN Solar Fuse line is UL and CSA recognized for PV systems.