Learn More
Grainger Solar Fuse 30A PVS-R Series 600VDC
Catalog No. 502752717
Encompass_Preferred
The Bussmann Solar Fuse 150 Amp is a 150 amp solar fuse designed for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass end caps and blade terminals. This fast-acting Class T fuse provides overcurrent protection for PV source circuits, PV output circuits, PV combiner boxes, and PV disconnects. It meets UL 2579 and IEC 60269 standards with a 10 x 38mm physical size. The 150 amp rating allows it to be used on systems up to 20kW.
Catalog No. 50-275-2717 Supplier Grainger Supplier No. 14L653
Add to Cart
Add to Cart
The Bussmann Solar Fuse 150 Amp is a 150 amp solar fuse designed for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass end caps and blade terminals. This fast-acting Class T fuse provides overcurrent protection for PV source circuits, PV output circuits, PV combiner boxes, and PV disconnects. It meets UL 2579 and IEC 60269 standards with a 10 x 38mm physical size. The 150 amp rating allows it to be used on systems up to 20kW.