Learn More
Grainger Solar Fuse 350A PVS-R Series 600VDC
Catalog No. 502749850
Encompass_Preferred
The BUSSMANN Solar Fuse 350 Amp is a high interrupting capacity fuse designed for photovoltaic systems. It has a voltage rating of 1000V DC and current rating of 350A. This Class J time-delay fuse provides overcurrent protection for conductors and equipment in PV systems. It features a ceramic body construction and blade-type terminals. The fuse measures 4-1/4"L x 1-5/8"W x 2-3/8"H. It can interrupt up to 300kA at 1000V DC. The 350A rating allows it to be used on systems up to 350kW. This UL and CSA listed fuse provides reliable overcurrent protection for solar applications.
Catalog No. 50-274-9850 Supplier Grainger Supplier No. 14L669
The BUSSMANN Solar Fuse 350 Amp is a high interrupting capacity fuse designed for photovoltaic systems. It has a voltage rating of 1000V DC and current rating of 350A. This Class J time-delay fuse provides overcurrent protection for conductors and equipment in PV systems. It features a ceramic body construction and blade-type terminals. The fuse measures 4-1/4"L x 1-5/8"W x 2-3/8"H. It can interrupt up to 300kA at 1000V DC. The 350A rating allows it to be used on systems up to 350kW. This UL and CSA listed fuse provides reliable overcurrent protection for solar applications.