Learn More
Grainger Solar Fuse 35A PVS-R Series 600VDC
Catalog No. 502749842
Encompass_Preferred
The Bussmann 14L654 Solar Fuse is a 35 amp, 600 volt DC photovoltaic fuse designed for PV systems. It has a compact cylindrical body made of ceramic and metal materials. This fast-acting fuse provides overload and short circuit protection for PV modules and balance-of-system components. It features a glass tube design for visual blown fuse indication. The 14L654 has a 35 amp current rating and is UL and CSA certified. With dimensions of 13.5mm diameter x 35mm length, this fuse provides reliable protection for solar applications.
Catalog No. 50-274-9842 Supplier Grainger Supplier No. 14L654
The Bussmann 14L654 Solar Fuse is a 35 amp, 600 volt DC photovoltaic fuse designed for PV systems. It has a compact cylindrical body made of ceramic and metal materials. This fast-acting fuse provides overload and short circuit protection for PV modules and balance-of-system components. It features a glass tube design for visual blown fuse indication. The 14L654 has a 35 amp current rating and is UL and CSA certified. With dimensions of 13.5mm diameter x 35mm length, this fuse provides reliable protection for solar applications.