Learn More
Grainger Solar Fuse 50A PVS-R Series 600VDC
Catalog No. 502749573
Encompass_Preferred
The BUSSMANN Solar Fuse 50A PVS-R Series 600VDC is a solar fuse designed for photovoltaic systems. It has a voltage rating of 600VDC and a current rating of 50A. This Class RK5 fuse provides overcurrent protection for conductors and components in a PV system. It features a ceramic body construction and blade-style terminals. The compact design allows for high density panel designs. This fuse meets UL recognition requirements and is listed under UL file number E19180. It can interrupt high DC fault currents up to 20kA. The 50A PVS-R fuse provides optimal protection for today's high power PV systems.
Catalog No. 50-274-9573 Supplier Grainger Supplier No. 14L656
The BUSSMANN Solar Fuse 50A PVS-R Series 600VDC is a solar fuse designed for photovoltaic systems. It has a voltage rating of 600VDC and a current rating of 50A. This Class RK5 fuse provides overcurrent protection for conductors and components in a PV system. It features a ceramic body construction and blade-style terminals. The compact design allows for high density panel designs. This fuse meets UL recognition requirements and is listed under UL file number E19180. It can interrupt high DC fault currents up to 20kA. The 50A PVS-R fuse provides optimal protection for today's high power PV systems.