Learn More
Grainger Solar Fuse 60A PVS-R Series 600VDC
Catalog No. 502749835
Encompass_Preferred
The Bussmann Solar Fuse 60 Amp is a UL listed, current limiting, high interrupting rating, time delay fuse designed for photovoltaic system overcurrent protection. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass endcaps and blade terminals. This 60 amp solar fuse provides overcurrent protection for conductors and components in a photovoltaic system. It has a time delay feature to allow harmless inrush currents during start up.
Catalog No. 50-274-9835 Supplier Grainger Supplier No. 14L657
The Bussmann Solar Fuse 60 Amp is a UL listed, current limiting, high interrupting rating, time delay fuse designed for photovoltaic system overcurrent protection. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass endcaps and blade terminals. This 60 amp solar fuse provides overcurrent protection for conductors and components in a photovoltaic system. It has a time delay feature to allow harmless inrush currents during start up.