Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PicoTurbine Solar Power Racer Eductaion Kit

Description
Students will learn how motors and automotive transmissions work. They will also expore how solar cells convert the energy from sunlight into electricity to power the car's motor.
Requires little to no previous knowledge of electronics to construct and operate. For 4th grade and up.
Kit includes:
- 1x Solar Cell
- 1x Metal body
- Gears, shafts, motor, and necessary parts
Specifications
Specifications
| Product Type | Kit, Solar Racer |
| Grade | 4+ |
| For Use With (Application) | teaching motors, transmission, solar energy and electricity concepts |
| Includes | Solar cell, metal body, gears, shafts, motor, and necessary parts. |
| Battery Type | Solar |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?