Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Solar-powered Hanging Scale
SDP

Catalog No. 22445590 Shop All Mopec Products
Change view
Click to view available options
Capacity:
30 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
22-445-590 30 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-445-590 Supplier Mopec™ Supplier No. BB067
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The BB067 Solar-powered Hanging Scale is easy to use, gives instantaneous weight readouts, and is completely solar powered, so you'll never have to worry about battery replacement.

  • Has the ability to zero, tare, bow, ring and pan
  • Digital speed for instantaneous weight readouts
  • Pinpoint accuracy to 0.01 kg (0.02 lb.)
  • Exclusive solar powered design for optimal efficiency
  • 30 lb. capacity for versatile weighing use
  • Easy to operate with no buttons to press

Specifications

Capacity 30 lb.
Display Type Digital
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.