Learn More
Peak Scientific Instruments Ltd Solaris 10 ELSD Nitrogen Generator Produces 10L/min of high purity Nitrogen
SDP
Catalog No. 502285707
Encompass_Preferred
Variable outlet flow, pressure and purity
UHP nitrogen for ELSD at up to 10 L/min, 100 psi and up to 99.5% purity
Compact benchtop form factor, discrete aesthetic styling
Colour indicative LED lighting for easy status awareness
Engineered on reliable and proven existing technology
Manufactured and tested to highest spec in UK, CE/FCC certified
12 month comprehensive on-site warranty
This item is not returnable. View return policy
Catalog No. 50-228-5707 Supplier Peak Scientific Instruments Ltd Supplier No. 650001
Add to Cart
Add to Cart
Request a Quote
Variable outlet flow, pressure and purity
UHP nitrogen for ELSD at up to 10 L/min, 100 psi and up to 99.5% purity
Compact benchtop form factor, discrete aesthetic styling
Colour indicative LED lighting for easy status awareness
Engineered on reliable and proven existing technology
Manufactured and tested to highest spec in UK, CE/FCC certified
12 month comprehensive on-site warranty