Learn More
Thermo Scientific™ Solaris 6000 Incubated and Refrigerated Stackable Shaker
Catalog No. 13642580 Shop All Thermo Scientific Products
Change view
Click to view available options
Model:
Incubated Stackable Shaker
Incubated Stackable Shaker with clamp starter kit
Incubated and Refrigerated Stackable Shaker
Incubated and Refrigerated Stackable Shaker with clamp starter kit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
13-642-580 Incubated Stackable Shaker
13-642-581 Incubated Stackable Shaker with clamp starter kit
13-642-582 Incubated and Refrigerated Stackable Shaker
13-642-579 Incubated and Refrigerated Stackable Shaker with clamp starter kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-642-580 Supplier Thermo Scientific™ Supplier No. SK6001
Add to Cart
Add to Cart
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
The Thermo Scientific™ Solaris 6000 Incubated and Refrigerated Stackable Shaker combines general purpose incubation with orbital shaking. The space saving stackable design and included shelving provides added versatility. Intuitive touch screen display, programmability, and refrigerant-free cooling.
The Thermo Scientific™ Solaris 6000 Incubated & Refrigerated Stackable Shaker combines general purpose incubation with orbital shaking. Peltier technology provides refrigerant-free cooling with quiet operation. The space saving stackable design and included shelving provides added versatility. Features intuitive touch screen display, programmability, on-unit performance monitoring, data export and multi-language support.
Thermo Scientific™ Solaris 6000 Incubated and Refrigerated Stackable Shakers are versatile general purpose laboratory equipment used for incubating and agitating samples. It combines the functions of an incubator and shaker, providing a temperature-controlled environment for cell culture, microbial growth, screening, molecular studies, and other applications.
The ergonomic stackable design enables increased sample capacity while minimizing footprint. Thoughtful design, including sealed electronics, rounded edges, and removable platform support ease of cleaning. Modern features include Peltier thermoelectric temperature control, touch screen user interface with programmability, performance logs, and data export. Shaking is supported by a high-performance triple eccentric drive mechanism, engineered to perform with a reduced noise level while supporting heavy loads with high reliability.
Excellent for a variety of applications
  • Units can be stacked two high on the floor to save valuable bench space
  • Two adjustable-height shelves provide additional incubated sample space
  • Large viewing window and internal light offer sample visibility
  • Optional inner chamber electrical outlet provides power for operating stirrers or rotators
  • Includes standard access port
Durable construction, reliable performance
  • Durable powder-coated, cold-rolled steel exterior
  • Precisely monitory and control chamber temperature over complete range with ±1°C accuracy at 37°C with PID temperature controller
  • Incubated and refrigerated models (Cat. No. SK6002) include thermoelectric temperature control
  • Overtemperature safety feature with independent thermostat provides additional backup by controlling heating if main temperature controller fails
  • Heavy-duty tri-eccentric drive mechanism, protected by overload sensor has a variable speed control from 15 to 525 rpm
  • Standard programmability allows storage of routines and setpoints for efficient use
  • Data monitoring alerts, alarms, and end of runs with on board graphing and run history
  • Spacious corrosion-resistant stainless steel chamber permits stacking an 18 x 18 platform to double the shaking capacity
  • Easy to read full color touch screen with multi language system
  • Includes 2 years labor, 5 years parts, and 10 years drive mechanism
TRUSTED_SUSTAINABILITY
Specifications
Orbit 25 mm
Load Bearing Capacity (Metric) 32 kg
Speed Range 15 to 525 rpm
Voltage 100/120/220/240 V
Model Incubated Stackable Shaker
Warranty and Service Offering 2 years labor, 5 years parts, and 10 years drive mechanism
Refrigerated No