Learn More
Peak Scientific Instruments Ltd Solaris XE Lab Nitrogen Generator Produces up to 35L/min of high purity N2
SDP
Catalog No. NC1703727
Encompass_Preferred
Nitrogen up to 35L/min, up to 116 psi and up to 99.5% purityVariable outlet flow, pressure and purityCompact, stackable* system maximizing the use of valuable laboratory floor or bench spaceColour indicative LED lighting for easy status awarenessManufactured and tested to highest spec in UK, CE/FCC/CSA certifiedSafer and more convenient than pressurized cylinders, dewars or bulk storage12 month on-site comprehensive warrantyBacked by [PEAK Protected] global on premise servicing and technical support
Catalog No. NC1703727 Supplier Peak Scientific Instruments Ltd Supplier No. 3300671
Add to Cart
Add to Cart
Request a Quote
Nitrogen up to 35L/min, up to 116 psi and up to 99.5% purityVariable outlet flow, pressure and purityCompact, stackable* system maximizing the use of valuable laboratory floor or bench spaceColour indicative LED lighting for easy status awarenessManufactured and tested to highest spec in UK, CE/FCC/CSA certifiedSafer and more convenient than pressurized cylinders, dewars or bulk storage12 month on-site comprehensive warrantyBacked by [PEAK Protected] global on premise servicing and technical support