Learn More
Peak Scientific Instruments Ltd Solaris XE Lab Nitrogen Generator for Waters Produces up to 35L/min of high purity N2 for Waters instruments
SDP
Catalog No. 5000804191
Encompass_Preferred
Nitrogen up to 35L/min, up to 116 psi and up to 99.5% purity
Variable outlet flow, pressure and purity
Compact, stackable* system maximizing the use of valuable laboratory floor or bench space
Color indicative LED lighting for easy status awareness
Manufactured and tested to highest spec in UK, CE/FCC certified
Safer and more convenient than pressurized cylinders, dewars or bulk storage
12 month on-site comprehensive warranty
Backed by [PEAK Protected] global on premise servicing and technical support
Catalog No. 50-008-04191 Supplier Peak Scientific Instruments Ltd Supplier No. 3300670
Nitrogen up to 35L/min, up to 116 psi and up to 99.5% purity
Variable outlet flow, pressure and purity
Compact, stackable* system maximizing the use of valuable laboratory floor or bench space
Color indicative LED lighting for easy status awareness
Manufactured and tested to highest spec in UK, CE/FCC certified
Safer and more convenient than pressurized cylinders, dewars or bulk storage
12 month on-site comprehensive warranty
Backed by [PEAK Protected] global on premise servicing and technical support