Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Solasodine | 126-17-0 | 10 MG
SDP

Catalog No. 5000298896
Encompass_Preferred

Solasodine is a steroidal alkaloid supplied as a research-grade solid for biochemical and pharmacological studies. The compound is provided in mg-scale laboratory packs for analytical use and is intended for research applications only.

  • Steroidal alkaloid research standard.
  • Solid powder form for analytical use.
  • CAS number 126-17-0 for unambiguous identification.
  • Molecular weight 413.64 g/mol.
  • Supplied in mg-scale laboratory packs suitable for research and analytical workflows.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-98896 Supplier Medchemexpress LLC Supplier No. HYN006810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solasodine is a steroidal alkaloid supplied as a research-grade solid for biochemical and pharmacological studies. The compound is provided in mg-scale laboratory packs for analytical use and is intended for research applications only.

  • Steroidal alkaloid research standard.
  • Solid powder form for analytical use.
  • CAS number 126-17-0 for unambiguous identification.
  • Molecular weight 413.64 g/mol.
  • Supplied in mg-scale laboratory packs suitable for research and analytical workflows.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.