Learn More
Medchemexpress LLC Solcitinib | 1206163-45-2 | 99.5% | 389.45 | 250 MG
SDP
Catalog No. 5000395174
Encompass_Preferred
Solcitinib is an orally active, competitive, potent, and selective JAK1 inhibitor with an IC50 of 9.8 nM. It exhibits 11-, 55-, and 23-fold selectivity over JAK2, JAK3, and TYK2, respectively, and is used in the research of moderate-to-severe plaque-type psoriasis.
  • Orally active
  • Competitive, potent, and selective JAK1 inhibitor
  • High selectivity over JAK2, JAK3, and TYK2
  • Used in research for moderate-to-severe plaque-type psoriasis
  • Available in solid and solution forms
Catalog No. 50-003-95174 Supplier Medchemexpress LLC Supplier No. 250MGHY16755
Solcitinib is an orally active, competitive, potent, and selective JAK1 inhibitor with an IC50 of 9.8 nM. It exhibits 11-, 55-, and 23-fold selectivity over JAK2, JAK3, and TYK2, respectively, and is used in the research of moderate-to-severe plaque-type psoriasis.
  • Orally active
  • Competitive, potent, and selective JAK1 inhibitor
  • High selectivity over JAK2, JAK3, and TYK2
  • Used in research for moderate-to-severe plaque-type psoriasis
  • Available in solid and solution forms