Learn More
Selleck Chemical LLC Solcitinib 5mg 1206163-45-2 GLPG0778, GSK2586184
SDP
Supplier:  Selleck Chemical LLC S59175MG
Encompass_Preferred
Solcitinib (GLPG0778, GSK2586184) is an inhibitor of JAK1 with an IC50 of 8-9 nM, and shows 11-, 55- and 23-fold selectivity over JAK2, JAK3 and TYK2, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8286