Learn More
NEWARK ELEMENT14 SOLDER WIRE
Catalog No. NC0680300
Encompass
SOLDER, WIRE, 180 C; External Diameter - Metric:0.5mm; External Diameter - Imperial:0.02 in.; Solder Alloy:60, 40 Sn, Pb; Melting Temperature:190 C; Weight:250g; Flux Type:-; Outer Diameter:0.5mm
Catalog No. NC0680300 Supplier NEWARK ELEMENT14 Supplier No. 15J5459
SOLDER, WIRE, 180 C; External Diameter - Metric:0.5mm; External Diameter - Imperial:0.02 in.; Solder Alloy:60, 40 Sn, Pb; Melting Temperature:190 C; Weight:250g; Flux Type:-; Outer Diameter:0.5mm