Learn More
MSC Weller Soldering Station, Digital, 85 W, 100-450 C, 1 ea
Catalog No. 501920245
Encompass
This digital soldering station operates at 120V and has an operating temperature range of 100 to 450 °C (200 to 850 °F). It features an 85 W power unit and is designed for efficient and reliable soldering, ensuring faster heat-up and recovery times for various applications.
  • Soldering iron is 70 W with an ergonomic handle and toolless tip change
  • ESD safe station, iron, and heat-resistant silicon cable
  • Standby mode and auto setback conserves energy and protects equipment
  • Password-protected to preserve settings
  • Uses ET soldering tips
  • Temperature stability and temperature lock for repeatable soldering results
  • Meets CE standards
Catalog No. 50-192-0245 Supplier MSC Supplier No. 46481404
This digital soldering station operates at 120V and has an operating temperature range of 100 to 450 °C (200 to 850 °F). It features an 85 W power unit and is designed for efficient and reliable soldering, ensuring faster heat-up and recovery times for various applications.
  • Soldering iron is 70 W with an ergonomic handle and toolless tip change
  • ESD safe station, iron, and heat-resistant silicon cable
  • Standby mode and auto setback conserves energy and protects equipment
  • Password-protected to preserve settings
  • Uses ET soldering tips
  • Temperature stability and temperature lock for repeatable soldering results
  • Meets CE standards