Learn More
Grainger Solenoid Coil Connector with Molded Wire Lead Type
Catalog No. NC1867039
Encompass_Preferred
This ARO solenoid coil connector helps you protect the solenoid valve electrical connections from humidity and moisture. Each connector is its own junction box so you don’t have to wire the solenoid to another box.
Catalog No. NC1867039 Supplier Grainger Supplier No. 4HN56
This ARO solenoid coil connector helps you protect the solenoid valve electrical connections from humidity and moisture. Each connector is its own junction box so you don’t have to wire the solenoid to another box.