Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Solenoid Valve 2Way. NC 12VDC

Catalog No. NC9538054 Shop All Thermo Scientific Products
Encompass_Preferred

Solenoid Valve 2Way. NC 12VDC.

Catalog No. NC9538054 Supplier Thermo Scientific Supplier No. 250118
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solenoid Valve 2Way. NC 12VDC.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.