Learn More
Julabo USA Inc Each, Cooling water solenoid for Forte HT30-M1 and HT60-ME.
SDP
Catalog No. 507534273
Encompass_Preferred
Bath Accessories, Julabo, Solenoid valve for cooling water (5°C to max 90°C). For use with Forte HT high temperature circulators only: HT30-M1 and HT60-M3. Automatically opens/closes cooling water flow when unti turned on/off.
Catalog No. 50-753-4273 Supplier Julabo USA Inc Supplier No. 8980704
Bath Accessories, Julabo, Solenoid valve for cooling water (5°C to max 90°C). For use with Forte HT high temperature circulators only: HT30-M1 and HT60-M3. Automatically opens/closes cooling water flow when unti turned on/off.