Learn More
Julabo USA Inc Each, Cooling water solenoid for HL/SL/MS/MX heating circulators
SDP
Catalog No. 507534179
Encompass_Preferred
Bath Accessories, Julabo, Solenoid valve for cooling water with Stakei connection. For use with HL-4, SL-6, SL-12, SL-26 heating circulators. Enables active cooling through the integrated cooling coil via tap water. Be sure to specify 115V or 230V when ordering. 8 mm Barb fittings.
Catalog No. 50-753-4179 Supplier Julabo USA Inc Supplier No. 8980703
Bath Accessories, Julabo, Solenoid valve for cooling water with Stakei connection. For use with HL-4, SL-6, SL-12, SL-26 heating circulators. Enables active cooling through the integrated cooling coil via tap water. Be sure to specify 115V or 230V when ordering. 8 mm Barb fittings.