Learn More
Julabo USA Inc Each, in-line solenoid valve set
SDP
Catalog No. 507534248
Encompass_Preferred
Bath Accessories, Julabo, Set of 2 solenoid valves (max. +100°C) to control in-line flow. Fittings: M16x1 male/female. Connects to Stakei outlet – make sure to specify 115V or 230V when ordering. Allows for fluid flow to application only when pump operates. Eliminates potential siphoning in the off state. For use with HL, SL, MS and MX circulators only.
Catalog No. 50-753-4248 Supplier Julabo USA Inc Supplier No. 8980701
Bath Accessories, Julabo, Set of 2 solenoid valves (max. +100°C) to control in-line flow. Fittings: M16x1 male/female. Connects to Stakei outlet – make sure to specify 115V or 230V when ordering. Allows for fluid flow to application only when pump operates. Eliminates potential siphoning in the off state. For use with HL, SL, MS and MX circulators only.