Learn More
Thermo Scientific SolEx HRPHS Polymer-Based SPE Cartridge | | Pack of 48
Catalog No. NC0855604 Shop All Thermo Scientific Products
Encompass
Obtain high recovery of polyphenols and related substances including humic acids and azo-containing substances such as azo dyes by using the Thermo Scientific Dionex SolEx HRPHS Polymer-Based SPE Cartridge The HRPHS stationary phase is a neutral resin comprised of a high-surface-area divinylbenzene-based particle grafted with a polyvinylpyrrolidone polymer This material has properties of a hydrophilic reversed-phase substance and also has specific high capacity for polyphenols The cartridge is dessigned for use with the Thermo Scientific Dionex AutoTrace 280 SPE system or other SPE instrumentation and vacuum manifolds
Catalog No. NC0855604 Supplier Thermo Scientific Supplier No. 088128
Obtain high recovery of polyphenols and related substances including humic acids and azo-containing substances such as azo dyes by using the Thermo Scientific Dionex SolEx HRPHS Polymer-Based SPE Cartridge The HRPHS stationary phase is a neutral resin comprised of a high-surface-area divinylbenzene-based particle grafted with a polyvinylpyrrolidone polymer This material has properties of a hydrophilic reversed-phase substance and also has specific high capacity for polyphenols The cartridge is dessigned for use with the Thermo Scientific Dionex AutoTrace 280 SPE system or other SPE instrumentation and vacuum manifolds